missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-acetylsalicylate, 98%, Thermo Scientific Chemicals

Product Code. 11479542
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Methyl 5-acetylsalicylate is used in food flavorings and preservatives. It is useful in the relief of headache and muscle and joint aches It is also effective in reducing fever, inflammation, and swelling and thus has been used for treatment of rheumatoid arthritis, rheumatic fever, and mild infection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 5-acetylsalicylate is used in food flavorings and preservatives. It is useful in the relief of headache and muscle and joint aches It is also effective in reducing fever, inflammation, and swelling and thus has been used for treatment of rheumatoid arthritis, rheumatic fever, and mild infection.

Solubility
Soluble in methanol.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16475-90-4
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00040924
InChI Key XLSMGNNWSRNTIQ-UHFFFAOYSA-N
Synonym methyl 5-acetylsalicylate, 5-acetylsalicylic acid methyl ester, 5-acetyl-2-hydroxybenzoic acid methyl ester, methyl 2-hydroxy-5-acetylbenzoate, benzoic acid, 5-acetyl-2-hydroxy-, methyl ester, 5-acetyl methyl salicylate, methyl 5-acethylsalicylate, pubchem22504, methyl5-acetylsalicylate, methyl-5-acetylsalicylate
PubChem CID 85444
IUPAC Name methyl 5-acetyl-2-hydroxybenzoate
SMILES CC(=O)C1=CC(=C(C=C1)O)C(=O)OC

Specifications

Melting Point 58°C to 62°C
Quantity 100 g
Beilstein 2643824
Solubility Information Soluble in methanol.
Formula Weight 194.19
Percent Purity 98%
Chemical Name or Material Methyl 5-acetylsalicylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'