Learn More
Methyl 5-bromopyridine-2-carboxylate, 98+%, Thermo Scientific Chemicals
41.31 € - 153.77 €
Chemical Identifiers
CAS | 29682-15-3 |
---|---|
Molecular Formula | C7H6BrNO2 |
Molecular Weight (g/mol) | 216.034 |
MDL Number | MFCD04112493 |
InChI Key | JEURNBCYNWNADN-UHFFFAOYSA-N |
Synonym | methyl 5-bromopicolinate, 5-bromopyridine-2-carboxylic acid methyl ester, methyl 5-bromo-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-pyridinecarboxylic acid methyl ester, 5-bromo-pyridine-2-carboxylic acid methyl ester, methyl5-bromopicolinate, pubchem10589, 5-bromopyridine-2-carboxylicacidmethylester, acmc-1cjr7Show More |
PubChem CID | 7016458 |
IUPAC Name | methyl 5-bromopyridine-2-carboxylate |
SMILES | COC(=O)C1=NC=C(C=C1)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11447096
|
Thermo Scientific Alfa Aesar
B25415.03 |
1 g | N/A | ||||||
11457096
|
Thermo Scientific Alfa Aesar
B25415.06 |
5 g |
153.77 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Methyl 5-bromopyridine-2-carboxylate is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 5-bromopyridine-2-carboxylate is used as an organic chemical synthesis intermediate.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
29682-15-3 | |
216.034 | |
JEURNBCYNWNADN-UHFFFAOYSA-N | |
7016458 | |
COC(=O)C1=NC=C(C=C1)Br |
C7H6BrNO2 | |
MFCD04112493 | |
methyl 5-bromopicolinate, 5-bromopyridine-2-carboxylic acid methyl ester, methyl 5-bromo-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-pyridinecarboxylic acid methyl ester, 5-bromo-pyridine-2-carboxylic acid methyl ester, methyl5-bromopicolinate, pubchem10589, 5-bromopyridine-2-carboxylicacidmethylester, acmc-1cjr7Show More | |
methyl 5-bromopyridine-2-carboxylate |
Specifications
29682-15-3 | |
C7H6BrNO2 | |
methyl 5-bromopicolinate, 5-bromopyridine-2-carboxylic acid methyl ester, methyl 5-bromo-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-pyridinecarboxylic acid methyl ester, 5-bromo-pyridine-2-carboxylic acid methyl ester, methyl5-bromopicolinate, pubchem10589, 5-bromopyridine-2-carboxylicacidmethylester, acmc-1cjr7 | |
JEURNBCYNWNADN-UHFFFAOYSA-N | |
methyl 5-bromopyridine-2-carboxylate | |
7016458 | |
≥98% |
99°C to 103°C | |
MFCD04112493 | |
Soluble in water. | |
COC(=O)C1=NC=C(C=C1)Br | |
216.034 | |
216.03 | |
Methyl 5-bromopyridine-2-carboxylate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.