missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 5-(chloromethyl)-2-furoate, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Quantity | 25 g |
Formula Weight | 174.58 |
Percent Purity | 95% |
Chemical Name or Material | Methyl 5-(chloromethyl)-2-furoate |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Methyl 5-(chloromethyl)-2-furoate, 95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction