missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-bromoindole-2-carboxylate, 97%, Thermo Scientific Chemicals

Product Code. 15409748
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Methyl 6-bromoindole-2-carboxylate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 6-bromoindole-2-carboxylate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in acetone.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372089-59-3
MDL Number MFCD03094942
Synonym ethyl 6-bromoindole-2-carboxylate, 6-bromoindole-2-carboxylic acid ethyl ester, 6-bromo-1h-indole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 6-bromo-, ethyl ester, 6-bromo-2-indolecarboxylic ethyl ester, 6-bromo-2-ethoxylcarbonyl-1h-indole, 6-bromo-2-carbethoxyindole, 6-bromo-2-carboethoxyindole, 1h-indole-2-carboxylicacid, 6-bromo-, ethyl ester
PubChem CID 7009496
IUPAC Name ethyl 6-bromo-1H-indole-2-carboxylate

Specifications

Color White
Molecular Formula C10H8BrNO2
Quantity 5 g
Solubility Information Soluble in acetone.
InChI Key BUMYRKNZGPXFEG-UHFFFAOYSA-N
SMILES COC(=O)C1=CC2=CC=C(Br)C=C2N1
Molecular Weight (g/mol) 254.08
Formula Weight 254.08
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 6-bromoindole-2-carboxylate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.