missing translation for 'onlineSavingsMsg'
Learn More
Methyl acetate, 99%, Thermo Scientific Chemicals Product Code.: 11373536

Methyl acetate, 99%, Thermo Scientific Chemicals

Product Code. 11373536
500 mL
Click to view available options
Quantity:
500 mL
1000 mL
2500 mL
This item is not returnable. View return policy

Product Code. 11373536

Brand: Thermo Scientific Alfa Aesar L14475.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methyl acetate is used as a solvent in glues, paints and nail polish removers. It is also used in adhesives, sealants and coatings. Further, it finds use as solvent in chemical reactions as well as used for extraction purpose. It is involved in the production of acetic anhydride by carbonylation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl acetate is used as a solvent in glues, paints and nail polish removers. It is also used in adhesives, sealants and coatings. Further, it finds use as solvent in chemical reactions as well as used for extraction purpose. It is involved in the production of acetic anhydride by carbonylation reaction.

Solubility
Miscible with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-20-9
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00008711
InChI Key KXKVLQRXCPHEJC-UHFFFAOYSA-N
Synonym tereton, devoton, acetic acid, methyl ester, methyl ethanoate, acetic acid methyl ester, methylacetat, acetate de methyle, methylacetaat, methyl acetic ester, octan metyluShow MoreShow Less
PubChem CID 6584
ChEBI CHEBI:77700
IUPAC Name methyl acetate
SMILES CC(=O)OC

Specifications

Melting Point -98°C
Density 0.934
Boiling Point 56°C to 58°C
Flash Point −10°C (14°F)
Odor Pleasant
Refractive Index 1.362
Quantity 500 mL
UN Number UN1231
Beilstein 1736662
Merck Index 14,6008
Solubility Information Miscible with water.
Formula Weight 74.08
Percent Purity 99%
Chemical Name or Material Methyl acetate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM03L006 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog NumberL14475.AP,L14475.AU,L14475.0F
Lot Number10218891 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL14475.AP,L14475.AU,L14475.0F
Lot Number10250937 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL14475.AP,L14475.AU,L14475.0F
Lot NumberN28K018 Certificate TypeCertificate of Analysis Date02/04/2024 Catalog NumberL14475.AP,L14475.AU,L14475.0F
Lot Number10238276 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL14475.AP,L14475.AU,L14475.0F
compliance-icons compliance-icons
Product Identifier
  • Methyl acetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Methyl acetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl acetate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.