missing translation for 'onlineSavingsMsg'
Learn More
Methyl aminomethanimidothioate hydroiodide, 95%, Thermo Scientific™ Product Code.: 10156932

Methyl aminomethanimidothioate hydroiodide, 95%, Thermo Scientific™

Product Code. 10156932
50 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10156932

Brand: Thermo Scientific Maybridge KM00619EE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 4338-95-8
Molecular Formula C2H7IN2S
Molecular Weight (g/mol) 218.06
MDL Number MFCD00035598
InChI Key LFXAECSQJSRSTP-UHFFFAOYSA-N
Synonym methyl aminomethanimidothioate hydroiodide, sulfiode, methylthiouronium iodide, methyl carbamimidothioate hydroiodide, s-methylthiuronium iodide, s-methylthiouronium iodide, s-methylisothiouronium iodide, 2-methyl-2-thiopseudourea hydroiodide, 2-methylisothiouronium iodide, s-methylisothiourea hydriodide
PubChem CID 197812
SMILES [H+].[I-].CSC(N)=N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 50 g
IUPAC Name hydrogen (methylsulfanyl)methanimidamide iodide
Formula Weight 218.06
Percent Purity 95%
Chemical Name or Material Methyl aminomethanimidothioate hydroiodide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl aminomethanimidothioate hydroiodide, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.