missing translation for 'onlineSavingsMsg'
Learn More

Methyl arachidate, 99%, Thermo Scientific Chemicals

Product Code. 15210846
Click to view available options
Quantity:
1 g
5 g
25 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-28-1
Molecular Formula C21H42O2
Molecular Weight (g/mol) 326.57
MDL Number MFCD00009014
InChI Key QGBRLVONZXHAKJ-UHFFFAOYSA-N
Synonym methyl arachidate, eicosanoic acid, methyl ester, arachidic acid methyl ester, methyl eicosanoate, methyl arachisate, arachidic acid methylester, eicosanoic acid methyl ester, methylarachidate, methyl aracidate
PubChem CID 14259
IUPAC Name methyl icosanoate
SMILES CCCCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 45.0°C to 48.0°C
Boiling Point 215.0°C to 216.0°C (10.0 mmHg)
Flash Point >110°C
Assay Percent Range 98.5 % min. (GC)
Packaging Glass Bottle
Linear Formula CH3(CH2)18COOCH3
Quantity 1 g
Formula Weight 326.56
Percent Purity 99%
Chemical Name or Material Methyl arachidate
Product Identifier
  • Methyl arachidate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'