missing translation for 'onlineSavingsMsg'
Learn More
Methyl benzoate, 99%, Thermo Scientific Chemicals Product Code.: 10081410

Methyl benzoate, 99%, Thermo Scientific Chemicals

Product Code. 10081410
2.5 L, Glass bottle
Click to view available options
Quantity:
500 mL
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10081410

Brand: Thermo Scientific Acros 126340025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-58-3
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
InChI Key QPJVMBTYPHYUOC-UHFFFAOYSA-N
Synonym methylbenzoate, benzoic acid, methyl ester, clorius, benzoic acid methyl ester, niobe oil, oil of niobe, methyl benzenecarboxylate, essence of niobe, oniobe oil, oxidate le
PubChem CID 7150
ChEBI CHEBI:72775
IUPAC Name methyl benzoate
SMILES COC(=O)C1=CC=CC=C1

Specifications

Acidity 1mg KOH/g max.
Melting Point -12.0°C
Color Colorless to Yellow
Density 1.0800g/mL
Boiling Point 198.0°C to 199.0°C
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CO2CH3
Refractive Index 1.5160 to 1.5180
Quantity 2.5 L
Beilstein 09,109
Merck Index 15,6098
Specific Gravity 1.08
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,methanol and ether
Viscosity 1.94 mPa.s (23°C)
Formula Weight 136.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl benzoate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0469417 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number126345000,126340025,126340100
Lot NumberA0460244 Certificate TypeCertificate of Analysis Date19/02/2024 Catalog Number126345000,12634010,126340025,126340100
Lot NumberA0455572 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number126345000,12634010,126340025,126340100
Lot NumberA0400674 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number126345000,12634010,126340025,126340100
Lot NumberA0405804 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number126345000,12634010,126340025,126340100
compliance-icons
Product Identifier
  • Methyl benzoate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl benzoate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.