missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl benzoylformate, 99%, Thermo Scientific Chemicals
26.40 € - 79.50 €
Chemical Identifiers
CAS | 15206-55-0 |
---|---|
Molecular Formula | C9H8O3 |
Molecular Weight (g/mol) | 164.16 |
MDL Number | MFCD00008443 |
InChI Key | YLHXLHGIAMFFBU-UHFFFAOYSA-N |
Synonym | methyl benzoylformate, methyl phenylglyoxylate, methyl oxophenylacetate, methyl phenylglyoxalate, benzoylformic acid methyl ester, methylbenzoylformate, phenylglyoxylic acid methyl ester, phenylglyoxylic acid, methyl ester, benzeneacetic acid, .alpha.-oxo-, methyl ester, unii-23f2045stg |
PubChem CID | 84835 |
ChEBI | CHEBI:84256 |
IUPAC Name | methyl 2-oxo-2-phenylacetate |
SMILES | COC(=O)C(=O)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10771745
|
Thermo Scientific Acros
126370050 |
5 g | Glass bottle |
26.40 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10685172
|
Thermo Scientific Acros
126370250 |
25 g | Glass bottle |
79.50 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15206-55-0 | |
164.16 | |
YLHXLHGIAMFFBU-UHFFFAOYSA-N | |
84835 | |
methyl 2-oxo-2-phenylacetate |
C9H8O3 | |
MFCD00008443 | |
methyl benzoylformate, methyl phenylglyoxylate, methyl oxophenylacetate, methyl phenylglyoxalate, benzoylformic acid methyl ester, methylbenzoylformate, phenylglyoxylic acid methyl ester, phenylglyoxylic acid, methyl ester, benzeneacetic acid, .alpha.-oxo-, methyl ester, unii-23f2045stg | |
CHEBI:84256 | |
COC(=O)C(=O)C1=CC=CC=C1 |
Specifications
15206-55-0 | |
Yellow | |
246°C to 248°C | |
Authentic | |
C9H8O3 | |
1.5250 to 1.527 | |
10, 657 | |
methyl benzoylformate, methyl phenylglyoxylate, methyl oxophenylacetate, methyl phenylglyoxalate, benzoylformic acid methyl ester, methylbenzoylformate, phenylglyoxylic acid methyl ester, phenylglyoxylic acid, methyl ester, benzeneacetic acid, .alpha.-oxo-, methyl ester, unii-23f2045stg | |
YLHXLHGIAMFFBU-UHFFFAOYSA-N | |
methyl 2-oxo-2-phenylacetate | |
84835 | |
164.16 | |
Liquid |
16°C | |
1.1500g/mL | |
77°C | |
98.5% min. (GC) | |
C6H5COCO2CH3 | |
MFCD00008443 | |
1.15 | |
Solubility in water: 2.02g/L (20°C) | |
COC(=O)C(=O)C1=CC=CC=C1 | |
164.16 | |
CHEBI:84256 | |
99% | |
Methyl benzoylformate, 99% |
Safety and Handling
missing translation for 'einecsNumber' : 239-263-3
missing translation for 'tsca' : TSCA
RUO – Research Use Only