missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl caffeate, Thermo Scientific Chemicals
Inhibitor of protein tyrosine kinase
91.10 € - 264.00 €
Chemical Identifiers
CAS | 3843-74-1 |
---|---|
Molecular Formula | C10H10O4 |
Molecular Weight (g/mol) | 194.186 |
MDL Number | MFCD00210468 |
InChI Key | OCNYGKNIVPVPPX-HWKANZROSA-N |
Synonym | methyl caffeate, methyl 3-3,4-dihydroxyphenyl acrylate, caffeic acid methyl ester, e-methyl 3-3,4-dihydroxyphenyl acrylate, methylcaffeate, methyl 3,4-dihydroxycinnamate, caffeic acid, methyl ester, unii-n79173b9hf, methyl 2e-3-3,4-dihydroxyphenyl prop-2-enoate, chembl17001 |
PubChem CID | 689075 |
ChEBI | CHEBI:6856 |
IUPAC Name | methyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate |
SMILES | COC(=O)C=CC1=CC(=C(C=C1)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15485949
|
Thermo Scientific Alfa Aesar
J63786.MD |
250 mg |
91.10 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15475949
|
Thermo Scientific Alfa Aesar
J63786.03 |
1 g |
264.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Inhibitor of protein tyrosine kinase
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3843-74-1 | |
194.186 | |
OCNYGKNIVPVPPX-HWKANZROSA-N | |
689075 | |
methyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate |
C10H10O4 | |
MFCD00210468 | |
methyl caffeate, methyl 3-3,4-dihydroxyphenyl acrylate, caffeic acid methyl ester, e-methyl 3-3,4-dihydroxyphenyl acrylate, methylcaffeate, methyl 3,4-dihydroxycinnamate, caffeic acid, methyl ester, unii-n79173b9hf, methyl 2e-3-3,4-dihydroxyphenyl prop-2-enoate, chembl17001 | |
CHEBI:6856 | |
COC(=O)C=CC1=CC(=C(C=C1)O)O |
Specifications
3843-74-1 | |
Yellow | |
MFCD00210468 | |
methyl caffeate, methyl 3-3,4-dihydroxyphenyl acrylate, caffeic acid methyl ester, e-methyl 3-3,4-dihydroxyphenyl acrylate, methylcaffeate, methyl 3,4-dihydroxycinnamate, caffeic acid, methyl ester, unii-n79173b9hf, methyl 2e-3-3,4-dihydroxyphenyl prop-2-enoate, chembl17001 | |
COC(=O)C=CC1=CC(=C(C=C1)O)O | |
194.186 | |
CHEBI:6856 | |
Powder |
158°C to 161°C | |
C10H10O4 | |
14,1635 | |
OCNYGKNIVPVPPX-HWKANZROSA-N | |
methyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate | |
689075 | |
194.19 | |
Methyl caffeate |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only