missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl carbamate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
100 g
500 g
Description
Methyl carbamate was used in the synthesis of protected aminocyclopropanes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl carbamate was used in the synthesis of protected aminocyclopropanes.
Solubility
Soluble in water(20g/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, metals, alkali metals.
Methyl carbamate was used in the synthesis of protected aminocyclopropanes.
Solubility
Soluble in water(20g/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, metals, alkali metals.
Specifications
Specifications
Melting Point | 54°C to 56°C |
Boiling Point | 176°C to 178°C |
Beilstein | 635779 |
Merck Index | 14,6036 |
Solubility Information | Soluble in water(20g/L). |
Formula Weight | 75.07 |
Percent Purity | 99% |
Density | 1.14 |
Quantity | 50 g |
Flash Point | >100°C (212°F) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'