missing translation for 'onlineSavingsMsg'
Learn More

Methyl cyclopropanecarboxylate, 98%

Produktkod. 11415806
Klicka för att se tillgängliga alternativ
Quantity:
25 g
100 g
Förpackningsstorlek:
100g
25g
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 11415806

Brand: Thermo Scientific Alfa Aesar B20853.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 2868-37-3 | C5H8O2 | 100.117 g/mol

It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products. Methyl cyclopropane carboxylate is used as a reagent in the synthesis of N’-thiazole and N-pyrazole based inhibitors of MAP kinase p38α.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products. Methyl cyclopropane carboxylate is used as a reagent in the synthesis of N’-thiazole and N-pyrazole based inhibitors of MAP kinase p38α.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents and ignition sources. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 2868-37-3
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00001280
InChI Key PKAHQJNJPDVTDP-UHFFFAOYSA-N
Synonym methyl cyclopropane carboxylate, cyclopropanecarboxylic acid, methyl ester, methylcyclopropanecarboxylate, cyclopropanecarboxylic acid methyl ester, methyl cyclopropylcarboxylate, unii-5uq53f585u, mcpc, pubchem11835, ksc490a7l, 1-methylcyclopropanecarboxylate
PubChem CID 76122
IUPAC Name methyl cyclopropanecarboxylate
SMILES COC(=O)C1CC1

Specifikationer

Density 0.983
Boiling Point 118°C to 119°C
Flash Point 22°C (71°F)
Refractive Index 1.418
Quantity 100 g
UN Number UN3272
Beilstein 2039388
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 100.12
Percent Purity 98%
Chemical Name or Material Methyl cyclopropanecarboxylate
Visa mer Visa mindre

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.