missing translation for 'onlineSavingsMsg'
Learn More
Methyl diethylphosphonoacetate, 97%, Thermo Scientific Chemicals Product Code.: 10168692

Methyl diethylphosphonoacetate, 97%, Thermo Scientific Chemicals

Product Code. 10168692
50 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10168692

Brand: Thermo Scientific Acros 169980500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1067-74-9
Molecular Formula C7H15O5P
Molecular Weight (g/mol) 210.17
MDL Number MFCD00009081
InChI Key CTSAXXHOGZNKJR-UHFFFAOYSA-N
Synonym methyl diethylphosphonoacetate, methyl 2-diethoxyphosphoryl acetate, acetic acid, diethoxyphosphinyl-, methyl ester, diethylmethylphosphonoacetate, methyl diethoxyphosphoryl acetate, diethyl carbomethoxymethylphosphonate, methyl 2-diethoxycarbonyl acetate, methyldiethylphosphonoacetate, diethyl methylphosphonoacetate, ksc489e0f
PubChem CID 66113
IUPAC Name methyl 2-diethoxyphosphorylacetate
SMILES CCOP(=O)(CC(=O)OC)OCC

Specifications

Color Colorless to Yellow
Density 1.1400g/mL
Boiling Point 127.0°C to 131.0°C (9.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5O)2P(O)CH2CO2CH3
Refractive Index 1.4320 to 1.4340
Quantity 50 g
Beilstein 04, II, 976
Specific Gravity 1.14
Formula Weight 210.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl diethylphosphonoacetate, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0398778 Certificate TypeCertificate of Analysis Date23/01/2023 Catalog Number169980500,169980100
Lot NumberA0358064 Certificate TypeCertificate of Analysis Date26/03/2021 Catalog Number169980500,169980100
Lot NumberA0332815 Certificate TypeCertificate of Analysis Date28/09/2020 Catalog Number169980500,169980100
compliance-icons
Product Identifier
  • Methyl diethylphosphonoacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl diethylphosphonoacetate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.