missing translation for 'onlineSavingsMsg'
Learn More
Methyl diethylphosphonoacetate, 97%, Thermo Scientific Chemicals Product Code.: 11473374

Methyl diethylphosphonoacetate, 97%, Thermo Scientific Chemicals

Product Code. 11473374
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11473374

Brand: Thermo Scientific Alfa Aesar A10644.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methyl diethylphosphonoacetate is used as a reagent in the preparation of allylic fluorides compounds and in regioselective Diels-Alder reactions. It also serves as a reactant in the preparation of substituted thiophenes and furans, which finds application in type 2 diabetes treatment. Further, it acts as a precursor in the synthesis of pyridone alkaloids and immunosuppressive cyclopentanediol derivatives. In addition to this, it is utilized in the modification of botulinum neurotoxin serotype A protease inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl diethylphosphonoacetate is used as a reagent in the preparation of allylic fluorides compounds and in regioselective Diels-Alder reactions. It also serves as a reactant in the preparation of substituted thiophenes and furans, which finds application in type 2 diabetes treatment. Further, it acts as a precursor in the synthesis of pyridone alkaloids and immunosuppressive cyclopentanediol derivatives. In addition to this, it is utilized in the modification of botulinum neurotoxin serotype A protease inhibitors.

Solubility
Miscible with tetrahydrofuran.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1067-74-9
Molecular Formula C7H15O5P
Molecular Weight (g/mol) 210.166
MDL Number MFCD00009081
InChI Key CTSAXXHOGZNKJR-UHFFFAOYSA-N
Synonym methyl diethylphosphonoacetate, methyl 2-diethoxyphosphoryl acetate, acetic acid, diethoxyphosphinyl-, methyl ester, diethylmethylphosphonoacetate, methyl diethoxyphosphoryl acetate, diethyl carbomethoxymethylphosphonate, methyl 2-diethoxycarbonyl acetate, methyldiethylphosphonoacetate, diethyl methylphosphonoacetate, ksc489e0f
PubChem CID 66113
IUPAC Name methyl 2-diethoxyphosphorylacetate
SMILES CCOP(=O)(CC(=O)OC)OCC

Specifications

Density 1.145
Boiling Point 126°C to 128°C (8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.432
Quantity 25 g
Beilstein 1782397
Solubility Information Miscible with tetrahydrofuran.
Formula Weight 210.17
Percent Purity 97%
Chemical Name or Material Methyl diethylphosphonoacetate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10100399 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberA10644.14
Lot Number10224588 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10644.14,A10644.36,A10644.22
Lot Number10159032 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10644.14,A10644.36,A10644.22
Lot NumberG28R027 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10644.14,A10644.36,A10644.22
Lot Number10183394 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10644.14,A10644.36,A10644.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl diethylphosphonoacetate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.