missing translation for 'onlineSavingsMsg'
Learn More
Methyl disulfide, 99%, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10790550

Methyl disulfide, 99%, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10790550
100 mL, AcroSeal™ Glass bottle
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 10790550

Brand: Thermo Scientific Acros 430821000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 624-92-0
Melting Point -85.0°C
Color Yellow
Density 1.0625g/mL
Boiling Point 108.0°C to 110.0°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 1% max. Methylmercaptan content (GC)
Packaging AcroSeal™ Glass bottle
Molecular Formula C2H6S2
Linear Formula (CH3)2S2
Refractive Index 1.5243 to 1.5263
MDL Number MFCD00008561
Quantity 100 mL
Specific Gravity 1.0625
Synonym dimethyl disulfide, methyl disulfide, dimethyldisulfide, disulfide, dimethyl, dmds, 2,3-dithiabutane, dimethyl disulphide, methyldisulfanyl methane, methyldisulfide, methyldithiomethane
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether,ethyl alcohol,ccl4,benzene
InChI Key WQOXQRCZOLPYPM-UHFFFAOYSA-N
SMILES CSSC
IUPAC Name (methyldisulfanyl)methane
Molecular Weight (g/mol) 94.19
PubChem CID 12232
ChEBI CHEBI:4608
Viscosity 0.620 mPa.s (20°C)
Formula Weight 94.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl disulfide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463097 Certificate TypeCertificate of Analysis Date20/05/2024 Catalog Number430821000
Lot NumberA0452222 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number430821000
Lot NumberA0423657 Certificate TypeCertificate of Analysis Date30/01/2023 Catalog Number430821000
Lot NumberA0403098 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number430821000
Lot NumberA0414696 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number430821000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methyl disulfide
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P311-Call a POISON CENTER/doctor/ .
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl disulfide, 99%, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.