missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl disulfide, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -85.0°C |
Color | Yellow |
Density | 1.0625g/mL |
Boiling Point | 108.0°C to 110.0°C |
Flash Point | 16°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 1% max. (Methylmercaptan content) (GC) |
Packaging | Glass bottle |
Linear Formula | (CH3)2S2 |
Refractive Index | 1.5243 to 1.5263 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0467274 | Certificate of Analysis | 01/11/2024 | 165590500,165592500,165590025,165590010 |
A0460609 | Certificate of Analysis | 27/02/2024 | 165590500,165592500,165590025,165590010 |
A0401628 | Certificate of Analysis | 15/12/2022 | 165590500,165592500,165590025,165590010 |
A0400684 | Certificate of Analysis | 15/12/2022 | 165590500,165592500,165590025,165590010 |
A0412959 | Certificate of Analysis | 13/12/2022 | 165590500,165592500,165590025,165590010 |
Safety and Handling




Signal Word
- Danger
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H370-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Methyl disulfide, 99%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction