missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Ethyl Ketone, HPLC Grade, J.T. Baker™
Click to view available options
Quantity:
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Description
- For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry
Specifications
Specifications
CAS | 78-93-3 |
Flash Point | −9.00°C (−48.20°F) |
Packaging | Narrow Mouth Amber Glass Bottle |
Molecular Formula | C4H8O |
MDL Number | MFCD00011648 |
Quantity | 4 L |
Specific Gravity | 1L = 0.81kg |
Synonym | 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon |
InChI Key | ZWEHNKRNPOVVGH-UHFFFAOYSA-N |
SMILES | CCC(C)=O |
Show More |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'