missing translation for 'onlineSavingsMsg'
Learn More

Methyl linolenate, 99%, Thermo Scientific Chemicals

Product Code. 10222532
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 301-00-8
Molecular Formula C19H32O2
Molecular Weight (g/mol) 292.46
MDL Number MFCD00135851
InChI Key DVWSXZIHSUZZKJ-YSTUJMKBSA-N
Synonym methyl linolenate, linolenic acid methyl ester, methyl alpha-linolenate, linolenic acid, methyl ester, unii-0s1ns923k6, alpha-linolenic acid methyl ester, methyl all-cis-9,12,15-octadecatrienoate, methyl 9z,12z,15z-octadeca-9,12,15-trienoate, 9,12,15-octadecatrienoic acid, methyl ester, z,z,z, methyl cis,cis,cis-octadec-9,12,15-trienoate
PubChem CID 5319706
IUPAC Name methyl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate
SMILES CC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)OC

Specifications

Color Colorless
Density 0.8950g/mL
Boiling Point 182.0°C (3.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2CH=CH)3(CH2)7CO2CH3
Refractive Index 1.4700 to 1.4720
Quantity 1 g
Beilstein 02, IV, 1782
Specific Gravity 0.895
Solubility Information Solubility in water: insoluble
Formula Weight 292.46
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl linolenate, 99%
Show More Show Less
Product Identifier
  • Methyl linolenate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'