Learn More
Methyl malonyl chloride, 97%, Thermo Scientific Chemicals
24.17 € - 177.00 €
Chemical Identifiers
CAS | 37517-81-0 |
---|---|
Molecular Formula | C4H5ClO3 |
Molecular Weight (g/mol) | 136.531 |
MDL Number | MFCD00013657 |
InChI Key | UTBCRHAMJFMIIR-UHFFFAOYSA-N |
PubChem CID | 123460 |
IUPAC Name | methyl 3-chloro-3-oxopropanoate |
SMILES | COC(=O)CC(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11400510
|
Thermo Scientific Alfa Aesar
020823.03 |
1 g |
24.17 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11357967
|
Thermo Scientific Alfa Aesar
020823.06 |
5 g |
46.33 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11490500
|
Thermo Scientific Alfa Aesar
020823.14 |
25 g |
177.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Methyl malonyl chloride is used as pharmaceutical intermediate. Used in anti-cancer drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
37517-81-0 | |
136.531 | |
UTBCRHAMJFMIIR-UHFFFAOYSA-N | |
methyl 3-chloro-3-oxopropanoate |
C4H5ClO3 | |
MFCD00013657 | |
123460 | |
COC(=O)CC(=O)Cl |
Specifications
37517-81-0 | |
1.27 g/mL | |
80°C (176°F) | |
1.4325 | |
UN3265 | |
methyl malonyl chloride, methyl 3-chloro-3-oxopropionate, methyl chloroformylacetate, propanoic acid, 3-chloro-3-oxo-, methyl ester, methyl-3-chloro-3-oxopropionate, methyl-3-chlor-3-oxopropanoat, methyl chlorocarbonylacetate, carbomethoxy acetyl chloride, methyl chloroformyl acetate, methyl3-chloro-3-oxopropionate | |
UTBCRHAMJFMIIR-UHFFFAOYSA-N | |
methyl 3-chloro-3-oxopropanoate | |
123460 | |
97% | |
Methyl malonyl chloride |
Colorless | |
58°C (12 mmHg) | |
C4H5ClO3 | |
MFCD00013657 | |
Moisture sensitive | |
Reacts with water. | |
COC(=O)CC(=O)Cl | |
136.531 | |
136.53 | |
Liquid |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 253-540-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.