missing translation for 'onlineSavingsMsg'
Learn More
Methyl methanesulfonate, 99%, Thermo Scientific Chemicals Product Code.: 15401278

Methyl methanesulfonate, 99%, Thermo Scientific Chemicals

Product Code. 15401278
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 15401278

Brand: Thermo Scientific Alfa Aesar H55120.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methyl methanesulfonate is a DNA adduct that adds methyl groups to Dan at 7-guanine, 3-guanine and 3-adenine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl methanesulfonate is a DNA adduct that adds methyl groups to Dan at 7-guanine, 3-guanine and 3-adenine.

Solubility
Very soluble in water.

Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-27-3
Molecular Formula C2H6O3S
Molecular Weight (g/mol) 110.127
MDL Number MFCD00007557
InChI Key MBABOKRGFJTBAE-UHFFFAOYSA-N
Synonym methyl mesylate, methanesulfonic acid methyl ester, methanesulfonic acid, methyl ester, methylmethane sulfonate, methylmethansulfonat, methyl methanesulphonate, as-dimethyl sulfite, as-dimethyl sulphite, methylmethanesulfonate, methyl methansulfonateShow MoreShow Less
PubChem CID 4156
ChEBI CHEBI:25255
IUPAC Name methyl methanesulfonate
SMILES COS(=O)(=O)C

Specifications

Color Colorless to Yellow
Density 1.3 g/mL
Boiling Point 202°C to 203°C
Flash Point 104°C (219°F)
Linear Formula CH3SO3CH3
Refractive Index 1.414
Quantity 5 g
UN Number UN2810
Beilstein 1098586
Merck Index 14,6097
Solubility Information Very soluble in water.
Formula Weight 110.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl methanesulfonate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0448640 Certificate TypeCertificate of Analysis Date20/09/2024 Catalog NumberH55120.06,H55120.14,H55120.22
Lot NumberR19I032 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberH55120.06,H55120.14,H55120.22
Lot NumberW12I005 Certificate TypeCertificate of Analysis Date16/09/2022 Catalog NumberH55120.06,H55120.14,H55120.22
Lot NumberP17I070 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberH55120.06,H55120.14,H55120.22
Lot NumberS30G052 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH55120.06,H55120.14,H55120.22
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methyl methanesulphonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H340-May cause genetic defects.
  • H351-Suspected of causing cancer.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Methyl methanesulphonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl methanesulfonate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.