missing translation for 'onlineSavingsMsg'
Learn More

Methyl nicotinate, 99%

Product Code. 10356480 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10356480

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 93-60-7 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-60-7
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00006388
InChI Key YNBADRVTZLEFNH-UHFFFAOYSA-N
Synonym methyl nicotinate, nikomet, methylnicotinate, nicotinic acid methyl ester, nicometh, methyl 3-pyridinecarboxylate, methyl-nicotinate, 3-pyridinecarboxylic acid, methyl ester, heat spray, nicotinic acid, methyl ester
PubChem CID 7151
IUPAC Name methyl pyridine-3-carboxylate
SMILES COC(=O)C1=CN=CC=C1

Specifications

Melting Point 38.0°C to 43.0°C
Color Brown to White
Boiling Point 209.0°C
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 500 g
Beilstein 22, 39
Merck Index 15, 6173
Solubility Information Solubility in water: negligible. Other solubilities: soluble in alcohol and benzene
Formula Weight 137.14
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Methyl nicotinate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl nicotinate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.