missing translation for 'onlineSavingsMsg'
Learn More

Methyl non-2-ynoate, ≥95%, Maybridge™

Product Code. 10466672
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 111-80-8
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.236
MDL Number MFCD00009547
InChI Key NTLJTUMJJWVCTL-UHFFFAOYSA-N
Synonym methyl 2-nonynoate, 2-nonynoic acid methyl ester, methyl octine carbonate, methyl octyne carbonate, 2-nonynoic acid, methyl ester, methyloctyne carboxylate, methyloctyne carbonate, methyl octin carbonate, octynecarboxylic acid, methyl ester, methyl octynecarboxylate
PubChem CID 8137
ChEBI CHEBI:51749
IUPAC Name methyl non-2-ynoate
SMILES CCCCCCC#CC(=O)OC

Specifications

CAS Min % 95.0
Density 0.9150g/mL
Assay Percent Range 95%+
Packaging Amber Glass Bottle
Linear Formula CH3(CH2)5Cidentical toCC(O)OCH3
Quantity 10 g
Formula Weight 168.23
Percent Purity 95%
Chemical Name or Material Methyl non-2-ynoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'