missing translation for 'onlineSavingsMsg'
Learn More

Methyl O,O'-bis(2,2,2-trifluoroethyl)phosphonoacetate, 90%, Thermo Scientific Chemicals

Product Code. 10160812
Click to view available options
5g; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Specifications

CAS 88738-78-7
Color Brown-Orange to Orange
Density 1.5040g/mL
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Molecular Formula C7H9F6O5P
Linear Formula CH3O2CCH2P(O)(OCH2CF3)2
Refractive Index 1.3700 to 1.3740
MDL Number MFCD00009901
Quantity 5 g
Fieser 12,320; 13,186
Specific Gravity 1.504
Synonym methyl 2-bis 2,2,2-trifluoroethoxy phosphoryl acetate, bis 2,2,2-trifluoroethyl methoxycarbonylmethyl phosphonate, methyl bis 2,2,2-trifluoroethyl phosphonoacetate, bis 2,2,2-trifluoroethyl phosphonoacetic acid methyl ester, methyl bis 2,2,2-trifluoroethoxy phosphoryl acetate, methyl p,p-bis 2,2,2-trifluoroethyl phosphonoacetate, acetic acid, bis 2,2,2-trifluoroethoxy phosphinyl-, methyl ester, acmc-209qx4
InChI Key PVSJXEDBEXYLML-UHFFFAOYSA-N
SMILES COC(=O)CP(=O)(OCC(F)(F)F)OCC(F)(F)F
IUPAC Name methyl 2-[bis(2,2,2-trifluoroethoxy)phosphoryl]acetate
Molecular Weight (g/mol) 318.11
PubChem CID 184894
Formula Weight 318.11
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Methyl O,O'-bis(2,2,2-trifluoroethyl)phosphonoacetate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl O,O'-bis(2,2,2-trifluoroethyl)phosphonoacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.