missing translation for 'onlineSavingsMsg'
Learn More

Methyl Orange Indicator, Reag. Ph. Eur., Honeywell Fluka™

Product Code. 15643820
Click to view available options
Quantity:
100 g
250 g
1.5 kg
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Indicator, Reag. Ph. Eur.

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 547-58-0
Molecular Formula C14H14N3NaO3S
Molecular Weight (g/mol) 327.334
MDL Number MFCD00007502
InChI Key STZCRXQWRGQSJD-UHFFFAOYSA-M
Synonym Helianthin
PubChem CID 23673835
IUPAC Name sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

Specifications

UN Number UN3143
Beilstein 4732884
Formula Weight 327.33g/mol
Packaging Glass Bottle
Suitability Complies for sensitivity
Visual Transition Interval 3.0 to 4.4
Color Orange
pH 3.0 to 4.4 (pink to yellow)
Quantity 1.5 kg
Physical Form Crystalline Powder
Chemical Name or Material Methyl Orange, Reag. Ph. Eur.
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'