missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Orange, Pure, Indicator Grade, Solid, C.I.13025, Fisher Chemical™
52.55 € - 104.00 €
Chemical Identifiers
CAS | 547-58-0 |
---|---|
Molecular Formula | C14H14N3NaO3S |
Molecular Weight (g/mol) | 327.334 |
MDL Number | 7502 |
InChI Key | STZCRXQWRGQSJD-UHFFFAOYSA-M |
PubChem CID | 23673835 |
IUPAC Name | sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate |
SMILES | CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+] |
Description
Chemical Identifiers
547-58-0 | |
327.334 | |
STZCRXQWRGQSJD-UHFFFAOYSA-M | |
sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate |
C14H14N3NaO3S | |
7502 | |
23673835 | |
CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+] |
Specifications
300°C | |
C14H14N3NaO3S | |
STZCRXQWRGQSJD-UHFFFAOYSA-M | |
sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate | |
23673835 | |
Glass Jar | |
Powder |
547-58-0 | |
7502 | |
CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+] | |
327.334 | |
327.32g/mol | |
Orange |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Methyl Orange
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /