missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Orange-Xylene Cyanol FF, Pure, Indicator Grade, Fisher Chemical
Click to view available options
Quantity:
500 mL
Packaging:
Amber glass bottle
Specifications
Specifications
MDL Number | 272193 |
Packaging | Amber glass bottle |
Color | Blue to Green |
Quantity | 500 mL |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Methyl orange-xylene cyanol FF
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P410-Protect from sunlight.
- MIXTURE LIST-Contains : Ethyl alcohol, Methyl alcohol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction