missing translation for 'onlineSavingsMsg'
Learn More

Methyl palmitate, 95%

Product Code. 10635871 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10635871

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-39-0 | C17H34O2 | 270.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-39-0
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.45
MDL Number MFCD00008994
InChI Key FLIACVVOZYBSBS-UHFFFAOYSA-N
Synonym methyl palmitate, palmitic acid methyl ester, hexadecanoic acid, methyl ester, palmitic acid, methyl ester, uniphat a60, metholene 2216, methyl n-hexadecanoate, hexadecanoic acid methyl ester, n-hexadecanoic acid methyl ester, unii-dpy8vcm98i
PubChem CID 8181
ChEBI CHEBI:69187
IUPAC Name methyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 28.0°C
Color Colorless
Density 0.85g/mL
Boiling Point 338.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)14CO2CH3
Quantity 100 g
Solubility Information Solubility in water: insoluble. Other solubilities: solubel in alcohol,ether
Formula Weight 270.45
Percent Purity 95%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material Methyl palmitate, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.