missing translation for 'onlineSavingsMsg'
Learn More

Methyl palmitoleate, 99%, analytical standard for GC, Thermo Scientific Chemicals

Product Code. 15540644
Click to view available options
Quantity:
1 g

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1120-25-8
Melting Point 0.5°C
Density 0.8800g/mL
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C17H32O2
MDL Number MFCD00042911
Quantity 1 g
Specific Gravity 0.88
Synonym methyl palmitoleate, z-methyl hexadec-9-enoate, methyl z-hexadec-9-enoate, methyl palmitoleinate, unii-35695qdb9f, methyl 9z-hexadec-9-enoate, 9-hexadecenoic acid, methyl ester, z, methyl cis-9-hexadecenoate, methyl z-9-hexadecenoate, methyl cis-hexadec-9-enoate
Solubility Information Solubility in water: insoluble.
InChI Key IZFGRAGOVZCUFB-HJWRWDBZSA-N
SMILES CCCCCC\C=C/CCCCCCCC(=O)OC
IUPAC Name methyl (Z)-hexadec-9-enoate
Molecular Weight (g/mol) 268.44
PubChem CID 643801
ChEBI CHEBI:84156
Formula Weight 268.43
Percent Purity 99%
Chemical Name or Material Methyl palmitoleate
Show More Show Less
Product Identifier
  • Methyl palmitoleate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.