missing translation for 'onlineSavingsMsg'
Learn More

Methyl perfluorooctanoate, 97%, Thermo Scientific™

Product Code. 11416706
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 376-27-2
Molecular Formula C9H3F15O2
Molecular Weight (g/mol) 428.097
MDL Number MFCD00039244
InChI Key XOCNYZFAMHDXJK-UHFFFAOYSA-N
Synonym methyl perfluorooctanoate, methyl pentadecafluorooctanoate, pentadecafluorooctanoic acid, methyl ester, methyl perfluorocaprylate, octanoic acid, pentadecafluoro-, methyl ester, perfluorooctanoic acid methyl ester, pentadecafluorooctanoic acid methyl ester, octanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-, methyl ester, acmc-1cndk, methylpentadecafluorooctanoate
PubChem CID 67824
IUPAC Name methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate
SMILES COC(=O)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Density 1.684
Boiling Point 158°C to 159°C
Flash Point 65°C (149°F)
Refractive Index 1.305
Quantity 25 g
Beilstein 1809817
Formula Weight 428.09
Percent Purity 97%
Chemical Name or Material Methyl perfluorooctanoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'