missing translation for 'onlineSavingsMsg'
Learn More
Methyl phenyl sulfoxide, 98+%, Thermo Scientific Chemicals Product Code.: 11417247

Methyl phenyl sulfoxide, 98+%, Thermo Scientific Chemicals

Product Code. 11417247
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11417247

Brand: Thermo Scientific Alfa Aesar A15009.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-82-4
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00002088
InChI Key JXTGICXCHWMCPM-UHFFFAOYSA-N
Synonym methyl phenyl sulfoxide, methylsulfinyl benzene, thioanisole s-oxide, phenyl methyl sulfoxide, sulfoxide, methyl phenyl, benzene, methylsulfinyl, methanesulfinylbenzene, methylphenylsulfoxide, methylsulphinyl benzene
PubChem CID 14516
IUPAC Name methylsulfinylbenzene
SMILES CS(=O)C1=CC=CC=C1

Specifications

Melting Point 26°C to 32°C
Boiling Point 93°C to 94°C (0.3 mmHg)
Flash Point 86°C (186°F)
Refractive Index 1.5775
Quantity 5 g
Beilstein 1904976
Sensitivity Hygroscopic
Formula Weight 140.2
Percent Purity ≥98%
Chemical Name or Material Methyl phenyl sulfoxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10219506 Certificate TypeCertificate of Analysis Date05/03/2024 Catalog NumberA15009.22,A15009.14,A15009.06,A1500901,A1500902
Lot Number10219599 Certificate TypeCertificate of Analysis Date05/03/2024 Catalog NumberA15009.22,A15009.14,A15009.06,A1500901,A1500902
Lot Number10219506 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15009.22,A15009.14,A15009.06
Lot Number10201745 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15009.22,A15009.14,A15009.06
Lot Number10219599 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15009.22,A15009.14,A15009.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl phenyl sulfoxide, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.