missing translation for 'onlineSavingsMsg'
Learn More
Methyl phenyl sulfoxide, 98+%, Thermo Scientific Chemicals Product Code.: 10653542

Methyl phenyl sulfoxide, 98+%, Thermo Scientific Chemicals

Product Code. 10653542
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10653542

Brand: Thermo Scientific Acros 352120250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-82-4
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.21
MDL Number MFCD00002088
InChI Key JXTGICXCHWMCPM-UHFFFAOYSA-N
Synonym methyl phenyl sulfoxide, methylsulfinyl benzene, thioanisole s-oxide, phenyl methyl sulfoxide, sulfoxide, methyl phenyl, benzene, methylsulfinyl, methanesulfinylbenzene, methylphenylsulfoxide, methylsulphinyl benzene
PubChem CID 14516
IUPAC Name methylsulfinylbenzene
SMILES CS(=O)C1=CC=CC=C1

Specifications

Melting Point 34°C to 36°C
Color White
Boiling Point 92°C to 94°C (0.3 mmHg)
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula C6H5SOCH3
Refractive Index 1.5775
Quantity 25 g
Solubility Information Solubility in water: insoluble (20°C)
Formula Weight 140.21
Percent Purity 98+%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Methyl phenyl sulfoxide, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0445488 Certificate TypeCertificate of Analysis Date19/09/2022 Catalog Number352120050,352120250
Lot NumberA0393971 Certificate TypeCertificate of Analysis Date14/09/2020 Catalog Number352120050,352120250
Product Identifier
  • Methyl phenyl sulfoxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl phenyl sulfoxide, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.