Learn More
Methyl pyruvate, 98%, Thermo Scientific Chemicals
Beschreibung
Methyl Pyruvate acts as a potent secretagogue used in the study of stimulus-secretion coupling. Also used in the preparation of quinoxalines as dual phoosphodiesterase 2/10 (PDE2/10) inhibitors. It increases free calcium ion concentration in the cytosol of pancreatic cells. It is used in the preparation of dimethyl 2,3-dimethylenebutanedioate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl Pyruvate acts as a potent secretagogue used in the study of stimulus-secretion coupling. Also used in the preparation of quinoxalines as dual phoosphodiesterase 2/10 (PDE2/10) inhibitors. It increases free calcium ion concentration in the cytosol of pancreatic cells. It is used in the preparation of dimethyl 2,3-dimethylenebutanedioate.
Solubility
Soluble in chloroform, methanol, ether, alcohol. Slightly soluble in water.
Notes
Store away from oxidizing agent, heat.

Spezifikation
Spezifikation
Density | 1.123 |
Boiling Point | 135°C to 137°C |
Flash Point | 39°C (102°F) |
Assay Percent Range | 98% |
Refractive Index | 1.404 |
Quantity | 25 g |
UN Number | UN3272 |
Beilstein | 1361953 |
Merck Index | 14,8021 |
Solubility Information | Soluble in chloroform,methanol,ether,alcohol. Slightly soluble in water. |
Mehr anzeigen |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10213808 | Certificate of Analysis | 30/08/2024 | A13966.14,A13966.36,A13966.22 |
10209482 | Certificate of Analysis | 22/11/2021 | A13966.14,A13966.36,A13966.22 |
10148520 | Certificate of Analysis | 22/11/2021 | A13966.14,A13966.36,A13966.22 |
10154336 | Certificate of Analysis | 22/11/2021 | A13966.14,A13966.36,A13966.22 |
10110243 | Certificate of Analysis | 22/11/2021 | A13966.14,A13966.36,A13966.22 |
Sicherheit und Handhabung



- Methyl pyruvat
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Methyl pyruvate
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.