missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Red, Pure, Water Soluble, Indicator, Fisher Chemical™
34.25 € - 167.00 €
Chemical Identifiers
CAS | 493-52-7 |
---|---|
Molecular Formula | C15H15N3O2 |
Molecular Weight (g/mol) | 269.304 |
MDL Number | 2425 |
InChI Key | CEQFOVLGLXCDCX-UHFFFAOYSA-N |
PubChem CID | 10303 |
IUPAC Name | 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid |
SMILES | CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O |
Description
Chemical Identifiers
493-52-7 | |
269.304 | |
CEQFOVLGLXCDCX-UHFFFAOYSA-N | |
2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid |
C15H15N3O2 | |
2425 | |
10303 | |
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O |
Specifications
178°C | |
C15H15N3O2 | |
CEQFOVLGLXCDCX-UHFFFAOYSA-N | |
2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid | |
10303 | |
Glass Jar | |
Solid |
493-52-7 | |
2425 | |
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O | |
269.304 | |
269.3g/mol | |
Red |