missing translation for 'onlineSavingsMsg'
Learn More
Methyl (S)-(-)-lactate, 97%, Thermo Scientific Chemicals Product Code.: 10573571

Methyl (S)-(-)-lactate, 97%, Thermo Scientific Chemicals

Product Code. 10573571
250 g, Glass bottle
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10573571

Brand: Thermo Scientific Acros 213062500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27871-49-4
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00064265
InChI Key LPEKGGXMPWTOCB-VKHMYHEASA-N
Synonym methyl s---lactate, methyl l-lactate, s-methyl lactate, --methyl l-lactate, s-lactic acid methyl ester, methyl 2s-2-hydroxypropanoate, --methyl lactate, methyl s-2-hydroxypropionate, propanoic acid, 2-hydroxy-, methyl ester, 2s, --lactic acid methyl ester
PubChem CID 94386
ChEBI CHEBI:83222
IUPAC Name methyl (2S)-2-hydroxypropanoate
SMILES COC(=O)[C@H](C)O

Specifications

Acidity 0.2% max. (as lactic acid)
Density 1.0920g/mL
Boiling Point 144.0°C to 145.0°C
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OH)CO2CH3
Refractive Index 1.4100 to 1.4150
Quantity 250 g
Beilstein 03, 264
Specific Gravity 1.092
Specific Rotation Condition − 8.50 (20.00°C neat)
Specific Rotation − 8.50
Solubility Information Solubility in water: miscible,hydrolyses. Other solubilities: miscible with most organic solvents,soluble in alcohol and ether
Viscosity 2.9 mPa.s (25°C)
Formula Weight 104.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl (S)-(-)-lactate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0448468 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number213060025,213062500,213060010
Lot NumberA0435548 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number213060025,213062500,213060010
Lot NumberA0398606 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number213060025,213062500,213060010
Lot NumberA0393293 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number213060025,213062500,213060010
Lot NumberA0396861 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number213060025,213062500,213060010
compliance-icons compliance-icons
Product Identifier
  • Methyl (S)-(-)-lactate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl (S)-(-)-lactate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.