Learn More
Methyl salicylate, 99%, Thermo Scientific Chemicals
30.55 € - 165.83 €
Chemical Identifiers
CAS | 119-36-8 |
---|---|
Molecular Formula | C8H8O3 |
Molecular Weight (g/mol) | 152.15 |
MDL Number | MFCD00002214 |
InChI Key | OSWPMRLSEDHDFF-UHFFFAOYSA-N |
Synonym | methyl salicylate, wintergreen oil, gaultheria oil, betula oil, teaberry oil, sweet birch oil, oil of wintergreen, analgit, spicewood oil, 2-hydroxybenzoic acid methyl ester |
PubChem CID | 4133 |
ChEBI | CHEBI:31832 |
IUPAC Name | methyl 2-hydroxybenzoate |
SMILES | COC(=O)C1=CC=CC=C1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10194700
|
Thermo Scientific Acros
220490050 |
5 mL | Glass bottle |
30.55 €
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10606052
|
Thermo Scientific Acros
220495000 |
500 mL | Glass bottle |
61.51 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10032970
|
Thermo Scientific Acros
220490010 |
1 L | Glass bottle |
97.18 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10687744
|
Thermo Scientific
220490025 |
2.5 L | Glass bottle |
165.83 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
119-36-8 | |
152.15 | |
OSWPMRLSEDHDFF-UHFFFAOYSA-N | |
4133 | |
methyl 2-hydroxybenzoate |
C8H8O3 | |
MFCD00002214 | |
methyl salicylate, wintergreen oil, gaultheria oil, betula oil, teaberry oil, sweet birch oil, oil of wintergreen, analgit, spicewood oil, 2-hydroxybenzoic acid methyl ester | |
CHEBI:31832 | |
COC(=O)C1=CC=CC=C1O |
Specifications
119-36-8 | |
Colorless to Yellow | |
222.0°C | |
Authentic | |
C8H8O3 | |
1.5350 to 1.5380 | |
10,7 | |
1.1825 | |
Solubility in water: 0.07 % (20°C). Other solubilities: soluble in glacial acetic acid, soluble in ether and chloroform, in ethanol 80 vol. %:1:2 v/v (20°C), diethyl phthalate:1:1 v/v (20°C), 1,2-propanediol:1/7 v/v (20°C) | |
COC(=O)C1=CC=CC=C1O | |
152.15 | |
CHEBI:31832 | |
99% | |
Methyl salicylate |
-8°C to 7°C | |
1.1825g/mL | |
110°C | |
98.5% min. (GC) | |
2-(HO)C6H4CO2CH3 | |
MFCD00002214 | |
15,6193 | |
methyl salicylate, wintergreen oil, gaultheria oil, betula oil, teaberry oil, sweet birch oil, oil of wintergreen, analgit, spicewood oil, 2-hydroxybenzoic acid methyl ester | |
OSWPMRLSEDHDFF-UHFFFAOYSA-N | |
methyl 2-hydroxybenzoate | |
4133 | |
152.15 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 204-317-7
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H361d-Suspected of damaging the unborn child.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only