missing translation for 'onlineSavingsMsg'
Learn More

Methyl stearate, mixture of homologs

Product Code. 10523421 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Glass Bottle
Plastic drum
Unit Size:
1kg
2.5kg
This item is not returnable. View return policy

Product Code. 10523421

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-61-8 | C19H38O2 | 298.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-61-8
Molecular Formula C19H38O2
Molecular Weight (g/mol) 298.51
MDL Number MFCD00009005
InChI Key HPEUJPJOZXNMSJ-UHFFFAOYSA-N
Synonym methyl stearate, octadecanoic acid, methyl ester, stearic acid methyl ester, metholene 2218, kemester 9718, methyl n-octadecanoate, stearic acid, methyl ester, kemester 9018, emery 2218, n-octadecanoic acid methyl ester
PubChem CID 8201
ChEBI CHEBI:69188
IUPAC Name methyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Acidity 1mg KOH/g max.
Melting Point 37.0°C to 39.0°C
Color Cream to White or Yellow
Density 0.8360g/mL
Boiling Point 215.0°C (15.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Linear Formula CH3(CH2)16CO2CH3
Quantity 1 kg
Saponification Value 186 to 192mg KOH/g
Merck Index 15,893
Specific Gravity 0.836
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether and alcohol
Formula Weight 298.51
Physical Form Paste
Chemical Name or Material Methyl stearate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.