missing translation for 'onlineSavingsMsg'
Learn More

Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals

Product Code. 10071470
10 mL, Glass bottle
Click to view available options
Quantity:
10 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-18-3
Molecular Formula C2H6S
Molecular Weight (g/mol) 62.13
MDL Number MFCD00008562
InChI Key QMMFVYPAHWMCMS-UHFFFAOYSA-N
Synonym dimethyl sulfide, methyl sulfide, methane, thiobis, dimethylsulfide, dimethylsulphide, methyl thioether, 2-thiapropane, dimethyl sulphide, dimethyl thioether, methylthiomethane
PubChem CID 1068
ChEBI CHEBI:17437
IUPAC Name methylsulfanylmethane
SMILES CSC

Specifications

Melting Point -98.0°C
Color Colorless
Density 0.8460g/mL
Boiling Point 38.0°C
Flash Point −48°C
Infrared Spectrum Authentic
Assay Percent Range 0.2% max. Methylmercaptan (GC)
Packaging Glass bottle
Linear Formula (CH3)2S
Refractive Index 1.4351
Quantity 10 mL
Beilstein 01, 288
Fieser 02,156; 04,190; 05,260; 15,88; 16,84; 17,198
Merck Index 15, 3281
Specific Gravity 0.846
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 62.13
Percent Purity 99+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Methyl sulfide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl sulfide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.