missing translation for 'onlineSavingsMsg'
Learn More

Methyl sulfoxide, 99.7%, pure, Thermo Scientific Chemicals

23.22 € - 367.83 €

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10021310
View Documents
Thermo Scientific Acros
127791000
100 mL Glass Bottle
23.22 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10571561
View Documents
Thermo Scientific Acros
1 L Glass Bottle
87.33 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10665942
View Documents
Thermo Scientific Acros
127790025
2.5 L Glass Bottle
103.52 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10716613
View Documents
Thermo Scientific Acros
127790050
5 L Plastic Drum
124.62 €
5 L
Please to purchase this item. Need a web account? Register with us today!
10764261
View Documents
Thermo Scientific Acros
127790250
25 L Plastic Drum
367.83 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

67-68-5
78.13
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
679
methylsulfinylmethane
C2H6OS
MFCD00002089
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
CHEBI:28262
CS(C)=O

Specifications

18.4°C
1.1000g/mL
87°C
Liquid
99.7%
C2H6OS
MFCD00002089
0.2% max.
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
methylsulfinylmethane
679
78.13
Pure
67-68-5
189.0°C
Authentic
1.4770 to 1.4800
1.1
(CH3)2SO
1.987 mPa.s (25°C)
01,296; 02,157; 03,119; 04,192; 05,263; 06,225; 07,133; 08,198; 09,189; 10,166; 12,212; 13,124; 15,146; 16,149; 17,127
Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, acetone, chloroform,, diethyl ether and aromatic hydrocarbons
CS(C)=O
78.13
CHEBI:28262
99.7%
Methyl sulfoxide, 99.7%
Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

missing translation for 'einecsNumber' : 200-664-3

missing translation for 'rtecsNumber' : PV6210000

missing translation for 'tsca' : TSCA

Product Identifier
  • Methyl sulfoxide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only