missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl sulfoxide, 99.8+%, extra pure, Thermo Scientific Chemicals
32.06 € - 833.15 €
Chemical Identifiers
CAS | 67-68-5 |
---|---|
Molecular Formula | C2H6OS |
Molecular Weight (g/mol) | 78.13 |
MDL Number | MFCD00002089 |
InChI Key | IAZDPXIOMUYVGZ-UHFFFAOYSA-N |
Synonym | dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexideShow More |
PubChem CID | 679 |
ChEBI | CHEBI:28262 |
IUPAC Name | methylsulfinylmethane |
SMILES | CS(C)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10043530
|
Thermo Scientific Acros
295522500 |
250 mL | Glass Bottle |
32.06 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10562841
|
Thermo Scientific Acros
295520010 |
1 L | Glass Bottle |
79.29 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10774923
|
Thermo Scientific Acros
295520025 |
2.5 L | Glass Bottle |
161.81 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10022923
|
Thermo Scientific Acros
|
25 L | Metal Drum |
833.15 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
67-68-5 | |
78.13 | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
679 | |
methylsulfinylmethane |
C2H6OS | |
MFCD00002089 | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexideShow More | |
CHEBI:28262 | |
CS(C)=O |
Specifications
18.4°C | |
0.04mg KOH/g max. | |
189.0°C | |
Liquid at 20°C | |
99.8+% | |
1.1 | |
(CH3)2SO | |
1.987 mPa.s (25°C) | |
0.1% max. | |
15, 3285 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, acetone, chloroform,, diethyl ether and aromatic hydrocarbons | |
CS(C)=O | |
78.13 | |
CHEBI:28262 | |
≥99.8% | |
Methyl sulfoxide |
67-68-5 | |
1.1000g/mL | |
87°C | |
1.4770 to 1.4800 | |
0.003% max. | |
C2H6OS | |
MFCD00002089 | |
01, IV, 1277 | |
01,296; 02,157; 03,119; 04,192; 05,263; 06,225; 07,133; 08,198; 09,189; 10,166; 12,212; 13,124; 15,146; 16,149; 17,127 | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
methylsulfinylmethane | |
679 | |
78.13 | |
Extra Pure |
Safety and Handling
Product Identifier
- Methyl sulfoxide
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Methyl sulfoxide, 99.8+%, extra pure, Thermo Scientific Chemicals