Methyl sulfoxide-d6, for NMR, contains 1 v/v% TMS, 99.9% atom D, Thermo Scientific Chemicals
Product Code.: 10696192
missing translation for 'structureSearchLink'
Print
Product Code. 10696192
10 mL , Glass Bottle
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mL
25 mL
50 mL
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
10696192
Brand: Thermo Scientific Acros
181150100
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
Chemical Identifiers
CAS
2206-27-1
Molecular Formula
C2H6OS
Molecular Weight (g/mol)
84.17
MDL Number
MFCD00002090
InChI Key
IAZDPXIOMUYVGZ-WFGJKAKNSA-N
Synonym
dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide Show More Show Less
PubChem CID
75151
IUPAC Name
trideuterio(trideuteriomethylsulfinyl)methane
SMILES
[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
Specifications
Physical Form
Liquid
Color
Colorless
Density
1.1900g/mL
Deuterium (D)
99.85% min.
Flash Point
88°C
Packaging
Glass Bottle
Specific Gravity
1.19
Viscosity
2.4 mPa.s (20°C)
Water
0.03% max. (Coulometric)
Quantity
10 mL
CAS Min %
99.85
Melting Point
20.2°C
Boiling Point
190.0°C
CAS Max %
100.0
Linear Formula
(CD3 )2 SO
Beilstein
01, IV, 1279
Fieser
13,124; 16,177
Formula Weight
84.18
Grade
NMR
Chemical Name or Material
Methyl sulfoxide-d6, 99.9 atom % D
Show More
Show Less
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number
Certificate Type
Certificate of Analysis
Certificate of Compliance
Certificate of Sterility/Irradiation
Certificate of Validation
Certificate of Guarantee
Certificate of Animal Origin
Certificate of Quality
Certificate of Processing
Declaration of Conformity
Search
5 results found
Lot Number
Certificate Type
Date
Catalog Number
Lot Number A0469684
Certificate Type Certificate of Analysis
Date 09/04/2025
Catalog Number 181150250,181150100,181150500
Lot Number A0466958
Certificate Type Certificate of Analysis
Date 25/01/2025
Catalog Number 181150250,181150100,181150500
Lot Number A0467889
Certificate Type Certificate of Analysis
Date 25/01/2025
Catalog Number 181150250,181150100,181150500
Lot Number A0465186
Certificate Type Certificate of Analysis
Date 02/09/2024
Catalog Number 181150250,181150100,181150500
Lot Number A0460270
Certificate Type Certificate of Analysis
Date 26/03/2024
Catalog Number 181150250,181150100,181150500
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Methyl sulfoxide-d6, for NMR, contains 1 v/v% TMS, 99.9% atom D, Thermo Scientific Chemicals >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.