missing translation for 'onlineSavingsMsg'
Learn More

Methyl-Tert-Butyl Ether, Extra Pure, SLR, Fisher Chemical™

Product Code. 10376330
2.5 L, Amber glass bottle
Click to view available options
Quantity:
500 mL
2.5 L
5 L
25 L
Packaging:
Amber glass bottle
HDPE Plastic Bottle
Metal can
Plastic Coated Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

C5H12O, CAS Number-1634-04-4, methyl t-butyl ether, 2-methyl-2-methoxypropane, methyl tert-butyl ether, propane, 2-methoxy-2-methyl, methyl-t-butyl ether, mtbe, methyl tertiary-butyl ether, methyl-tert-butyl ether, tert-butyl methyl ether, t-butyl methyl ether, 5L, 54 deg.C, CHEBI:27642, Colorless

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number 8812
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Melting Point -110°C
Color Colorless
Boiling Point 54°C
Packaging Amber glass bottle
Quantity 2.5 L
Formula Weight 88.15g/mol
Vapor Pressure 268mbar at 20°C
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl methyl ether
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl-Tert-Butyl Ether, Extra Pure, SLR, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.