Learn More
Methyl-Tert-Butyl Ether, for HPLC, Fisher Chemical™
C5H12O, CAS Number-1634-04-4, mtbe, methyl tert-butyl ether, methyl t-butyl ether, 2-methyl-2-methoxypropane, methyl-tert-butyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-t-butyl ether, tert-butyl methyl ether, t-butyl methyl ether, 2.5L, 54 deg.C, CHEBI:27642, Colorless
Brand: Fisher Chemical M/4496/17
227.35 EUR valid until 2024-12-31
Use promo code "21665" to get your promotional price.
Description
C5H12O, CAS Number-1634-04-4, mtbe, methyl tert-butyl ether, methyl t-butyl ether, 2-methyl-2-methoxypropane, methyl-tert-butyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-t-butyl ether, tert-butyl methyl ether, t-butyl methyl ether, 2.5L, 54 deg.C, CHEBI:27642, Colorless
Specifications
1634-04-4 | |
Colorless | |
Amber glass bottle | |
8812 | |
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane | |
CC(C)(C)OC | |
88.15 | |
CHEBI:27642 | |
268mbar at 20°C |
-110°C | |
54°C | |
C5H12O | |
2.5 L | |
BZLVMXJERCGZMT-UHFFFAOYSA-N | |
2-methoxy-2-methylpropane | |
15413 | |
88.15g/mol | |
Liquid |