missing translation for 'onlineSavingsMsg'
Learn More

Methyl vanillate, 99%, Thermo Scientific Chemicals

30.25 € - 299.49 €

Chemical Identifiers

CAS 3943-74-6
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00008438
InChI Key BVWTXUYLKBHMOX-UHFFFAOYSA-N
Synonym methyl vanillate, methyl 3-methoxy-4-hydroxybenzoate, vanillic acid, methyl ester, 4-hydroxy-3-methoxybenzoic acid methyl ester, methylvanillate, benzoic acid, 4-hydroxy-3-methoxy-, methyl ester, vanillic acid methyl ester, unii-2hxg8qso3d, 2hxg8qso3d, vxx
PubChem CID 19844
ChEBI CHEBI:46477
IUPAC Name methyl 4-hydroxy-3-methoxybenzoate
SMILES COC1=C(C=CC(=C1)C(=O)OC)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10136130
View Documents
Thermo Scientific Acros
160300050
5 g Glass bottle
30.25 €
5g
Please to purchase this item. Need a web account? Register with us today!
10002050
View Documents
Thermo Scientific Acros
160300250
25 g Glass bottle
120.60 €
25g
Please to purchase this item. Need a web account? Register with us today!
10310214
View Documents
Thermo Scientific Acros
160301000
100 g Glass bottle
299.49 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3943-74-6
182.18
BVWTXUYLKBHMOX-UHFFFAOYSA-N
19844
methyl 4-hydroxy-3-methoxybenzoate
C9H10O4
MFCD00008438
methyl vanillate, methyl 3-methoxy-4-hydroxybenzoate, vanillic acid, methyl ester, 4-hydroxy-3-methoxybenzoic acid methyl ester, methylvanillate, benzoic acid, 4-hydroxy-3-methoxy-, methyl ester, vanillic acid methyl ester, unii-2hxg8qso3d, 2hxg8qso3d, vxx
CHEBI:46477
COC1=C(C=CC(=C1)C(=O)OC)O

Specifications

3943-74-6
Beige to Yellow
Authentic
C9H10O4
MFCD00008438
methyl vanillate, methyl 3-methoxy-4-hydroxybenzoate, vanillic acid, methyl ester, 4-hydroxy-3-methoxybenzoic acid methyl ester, methylvanillate, benzoic acid, 4-hydroxy-3-methoxy-, methyl ester, vanillic acid methyl ester, unii-2hxg8qso3d, 2hxg8qso3d, vxx
COC1=C(C=CC(=C1)C(=O)OC)O
182.18
CHEBI:46477
99%
Methyl vanillate
64.0°C to 67.0°C
285.0°C to 287.0°C
98.5% min. (GC)
HOC6H3(OCH3)CO2CH3
10, 396
BVWTXUYLKBHMOX-UHFFFAOYSA-N
methyl 4-hydroxy-3-methoxybenzoate
19844
182.18
Powder or Granules
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 223-525-9

Product Identifier
  • Methyl vanillate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only