missing translation for 'onlineSavingsMsg'
Learn More

Methyl Viologen hydrate, 98%, Thermo Scientific Chemicals

74.57 € - 272.36 €

Chemical Identifiers

CAS 1910-42-5
Molecular Formula C12H14Cl2N2
Molecular Weight (g/mol) 257.16
MDL Number MFCD00150001
InChI Key FIKAKWIAUPDISJ-UHFFFAOYSA-L
Synonym Paraquat, dichloride
PubChem CID 15938
ChEBI CHEBI:28786
IUPAC Name 1-methyl-4-(1-methylpyridin-1-ium-4-yl)pyridin-1-ium;dichloride
SMILES [Cl-].[Cl-].C[N+]1=CC=C(C=C1)C1=CC=[N+](C)C=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10358030
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Acros
227320010
1 g Glass bottle
74.57 €
1g
Please to purchase this item. Need a web account? Register with us today!
10114860
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Acros
227320050
5 g Glass bottle
272.36 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1910-42-5
257.16
FIKAKWIAUPDISJ-UHFFFAOYSA-L
15938
1-methyl-4-(1-methylpyridin-1-ium-4-yl)pyridin-1-ium;dichloride
C12H14Cl2N2
MFCD00150001
Paraquat, dichloride
CHEBI:28786
[Cl-].[Cl-].C[N+]1=CC=C(C=C1)C1=CC=[N+](C)C=C1

Specifications

>300.0°C
98%
MFCD00150001
Solubility in water: very soluble in water.
Authentic
1-methyl-4-(1-methylpyridin-1-ium-4-yl)pyridin-1-ium;dichloride
15938
257.15
Beige or Brown to Pink
Methyl Viologen hydrate, 98%
1910-42-5
C12H14Cl2N2
Paraquat, dichloride
FIKAKWIAUPDISJ-UHFFFAOYSA-L
[Cl-].[Cl-].C[N+]1=CC=C(C=C1)C1=CC=[N+](C)C=C1
257.16
CHEBI:28786
98%
Crystalline Powder or Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Fatal in contact with skin.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

missing translation for 'einecsNumber' : 217-615-7

missing translation for 'rtecsNumber' : DW2275000

Product Identifier
  • Methyl Viologen hydrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.

SDS
Documents

Documents

Certificates

RUO – Research Use Only