missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylamine, Thermo Scientific Chemicals
101.51 € - 626.12 €
Specifications
Chemical Name or Material | Methylamine |
---|---|
CAS | 74-89-5, 109-99-9 |
Molecular Formula | CH5N |
Linear Formula | CH3NH2 |
Specific Gravity | 0.861 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10374552
|
Thermo Scientific Acros
368491000 |
100 mL |
101.51 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10165683
|
Thermo Scientific Acros
368498000 |
800 mL |
626.12 €
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methylamine | |
CH5N | |
0.861 | |
BAVYZALUXZFZLV-UHFFFAOYSA-N | |
methanamine | |
CHEBI:16830 | |
Pure |
74-89-5, 109-99-9 | |
CH3NH2 | |
methylamine, aminomethane, monomethylamine, carbinamine, mercurialin, n-methylamine, methylaminen, metilamine, metyloamina, anhydrous methylamine | |
CN | |
6329 | |
31.06 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Methylamine, 2M solution in THF
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Methylamine, Thermo Scientific Chemicals