missing translation for 'onlineSavingsMsg'
Learn More
Methylboronic acid, 97%, Thermo Scientific Chemicals Product Code.: 10664365

Methylboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 10664365
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10664365

Brand: Thermo Scientific Acros 344670010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13061-96-6
Molecular Formula CH5BO2
Molecular Weight (g/mol) 59.86
MDL Number MFCD00002105
InChI Key KTMKRRPZPWUYKK-UHFFFAOYSA-N
Synonym methaneboronic acid, methyl boronic acid, dihydroxymethylborane, boronic acid, methyl, unii-8z1me41sch, methane boronic acid, 8z1me41sch, methylboronicacid, methyboronic acid, methyl-boronic acid
PubChem CID 139377
IUPAC Name methylboronic acid
SMILES CB(O)O

Specifications

Melting Point 88.0°C to 90.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3B(OH)2
Quantity 1 g
Solubility Information Solubility in water: soluble in water
Formula Weight 59.86
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Methylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • Methylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methylboronic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.