missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyle1H-benzimidazole-5-carboxylate, 97%, Thermo Scientific™
11.30 € - 83.70 €
Chemical Identifiers
CAS | 26663-77-4 |
---|---|
Molecular Formula | C9H8N2O2 |
Molecular Weight (g/mol) | 176.175 |
MDL Number | MFCD03407310 |
InChI Key | WJHHIVYNOVTVGY-UHFFFAOYSA-N |
Synonym | methyl benzimidazole-5-carboxylate, methyl 1h-benzimidazole-5-carboxylate, methyl 1h-benzo d imidazole-5-carboxylate, methyl 1h-1,3-benzodiazole-5-carboxylate, 1h-benzimidazole-5-carboxylicacidmethylester, 1h-benzimidazole-5-carboxylic acid methyl ester, methyl 1h-benzo d imidazole-6-carboxylate, benzimidazole-5-carboxylic acid methyl ester, 1h-benzoimidazole-5-carboxylic acid methyl ester, 1h-benzimidazole-5-carboxylic acid, methyl ester |
PubChem CID | 2779733 |
IUPAC Name | methyl 3H-benzimidazole-5-carboxylate |
SMILES | COC(=O)C1=CC2=C(C=C1)N=CN2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10213032
|
Thermo Scientific Maybridge
MO00298CB |
250 mg | Amber Glass Bottle |
24.20 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10171312
|
Thermo Scientific Maybridge
MO00298DA |
1 g | Amber Glass Bottle |
11.30 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10317074
|
Thermo Scientific Maybridge
MO00298EA |
10 g | Amber Glass Bottle |
83.70 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
26663-77-4 | |
176.175 | |
WJHHIVYNOVTVGY-UHFFFAOYSA-N | |
2779733 | |
COC(=O)C1=CC2=C(C=C1)N=CN2 |
C9H8N2O2 | |
MFCD03407310 | |
methyl benzimidazole-5-carboxylate, methyl 1h-benzimidazole-5-carboxylate, methyl 1h-benzo d imidazole-5-carboxylate, methyl 1h-1,3-benzodiazole-5-carboxylate, 1h-benzimidazole-5-carboxylicacidmethylester, 1h-benzimidazole-5-carboxylic acid methyl ester, methyl 1h-benzo d imidazole-6-carboxylate, benzimidazole-5-carboxylic acid methyl ester, 1h-benzoimidazole-5-carboxylic acid methyl ester, 1h-benzimidazole-5-carboxylic acid, methyl ester | |
methyl 3H-benzimidazole-5-carboxylate |
Specifications
26663-77-4 | |
97% | |
MFCD03407310 | |
WJHHIVYNOVTVGY-UHFFFAOYSA-N | |
methyl 3H-benzimidazole-5-carboxylate | |
2779733 | |
97% |
97.0 | |
C9H8N2O2 | |
methyl benzimidazole-5-carboxylate, methyl 1h-benzimidazole-5-carboxylate, methyl 1h-benzo d imidazole-5-carboxylate, methyl 1h-1,3-benzodiazole-5-carboxylate, 1h-benzimidazole-5-carboxylicacidmethylester, 1h-benzimidazole-5-carboxylic acid methyl ester, methyl 1h-benzo d imidazole-6-carboxylate, benzimidazole-5-carboxylic acid methyl ester, 1h-benzoimidazole-5-carboxylic acid methyl ester, 1h-benzimidazole-5-carboxylic acid, methyl ester | |
COC(=O)C1=CC2=C(C=C1)N=CN2 | |
176.175 | |
176.17 | |
Methyl 1H-benzimidazole-5-carboxylate |