missing translation for 'onlineSavingsMsg'
Learn More

Methyle2-(acetylamino)-5-nitrobenzoate, 97%, Thermo Scientific™

Product Code. 10180932
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 5409-45-0
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H10N2O5
MDL Number MFCD00157624
Quantity 10 g
Synonym methyl 2-acetylamino-5-nitrobenzoate, acmc-20akiu, maybridge1_006182, methyl 2-acetamido-5-nitrobenzoate, methyl2-acetylamino-5-nitrobenzoate, 2-acetyl-amino-5-nitro-benzoic acid methyl ester, benzoic acid,2-acetylamino-5-nitro-, methyl ester
InChI Key HAKPLTBAUCAFMV-UHFFFAOYSA-N
SMILES CC(=O)NC1=C(C=C(C=C1)[N+](=O)[O-])C(=O)OC
IUPAC Name methyl 2-acetamido-5-nitrobenzoate
Molecular Weight (g/mol) 238.199
PubChem CID 224172
Formula Weight 238.2
Percent Purity 97%
Chemical Name or Material Methyl 2-(acetylamino)-5-nitrobenzoate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'