Learn More
Methyl 2-aminopyridine-4-carboxylate, 98%, Thermo Scientific Chemicals
46.90 € - 139.00 €
Chemical Identifiers
CAS | 6937-03-7 |
---|---|
Molecular Formula | C7H8N2O2 |
Molecular Weight (g/mol) | 152.15 |
MDL Number | MFCD04039316 |
InChI Key | SVWWNEYBEFASMP-UHFFFAOYSA-N |
IUPAC Name | methyl 2-aminopyridine-4-carboxylate |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6937-03-7 | |
152.15 | |
SVWWNEYBEFASMP-UHFFFAOYSA-N |
C7H8N2O2 | |
MFCD04039316 | |
methyl 2-aminopyridine-4-carboxylate |
Specifications
6937-03-7 | |
White to Brown | |
97.5% min. (GC) | |
MFCD04039316 | |
SVWWNEYBEFASMP-UHFFFAOYSA-N | |
methyl 2-aminopyridine-4-carboxylate | |
236984 | |
98% | |
Methyl 2-aminopyridine-4-carboxylate |
145.0°C to 148.0°C | |
Authentic | |
C7H8N2O2 | |
methyl 2-aminoisonicotinate, 2-aminopyridine-4-carboxylic acid methyl ester, 2-aminoisonicotinic acid methyl ester, 4-methoxycarbonyl pyridin-2-amine, 4-pyridinecarboxylic acid, 2-amino-, methyl ester, methyl 2-amino-4-pyridinecarboxylate, methyl-2-aminopyridine-4-carboxylate, 2-amino-4-pyridinecarboxylic acid methylester, 2-amino-4-pyridinecarboxylic acid methyl ester, methyl-2-aminoisonicotinat | |
COC(=O)C1=CC(=NC=C1)N | |
152.15 | |
152.15 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash
GHS Signal Word: Warning
- Methyl 2-aminopyridine-4-carboxylate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only