missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyle3-hydroxybutanoate, 97%, Thermo Scientific™
30.60 € - 172.00 €
Chemical Identifiers
CAS | 3976-69-0 |
---|---|
Molecular Formula | C5H10O3 |
Molecular Weight (g/mol) | 118.132 |
MDL Number | MFCD00063289 |
InChI Key | LDLDJEAVRNAEBW-SCSAIBSYSA-N |
Synonym | methyl r---3-hydroxybutyrate, r-methyl 3-hydroxybutanoate, methyl 3r-3-hydroxybutanoate, methyl r-3-hydroxybutyrate, r-3-hydroxybutyric acid methyl ester, methyl r-3-hydroxybutanoate, r---3-hydroxybutyric acid methyl ester, r---methyl 3-hydroxybutyrate, r---3-hydroxy-n-butyric acid methyl ester |
PubChem CID | 2724279 |
IUPAC Name | methyl (3R)-3-hydroxybutanoate |
SMILES | CC(CC(=O)OC)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10778835
|
Thermo Scientific Maybridge
SB01506EA |
10 g |
30.60 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10526334
|
Thermo Scientific Maybridge
SB01506EB |
25 g |
62.80 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10668545
|
Thermo Scientific Maybridge
SB01506FA |
100 g |
172.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
3976-69-0 | |
118.132 | |
LDLDJEAVRNAEBW-SCSAIBSYSA-N | |
2724279 | |
CC(CC(=O)OC)O |
C5H10O3 | |
MFCD00063289 | |
methyl r---3-hydroxybutyrate, r-methyl 3-hydroxybutanoate, methyl 3r-3-hydroxybutanoate, methyl r-3-hydroxybutyrate, r-3-hydroxybutyric acid methyl ester, methyl r-3-hydroxybutanoate, r---3-hydroxybutyric acid methyl ester, r---methyl 3-hydroxybutyrate, r---3-hydroxy-n-butyric acid methyl ester | |
methyl (3R)-3-hydroxybutanoate |
Specifications
3976-69-0 | |
1.0500g/mL | |
Amber glass bottle | |
CH3CH(OH)CH2CO2CH3 | |
- 21.50 (20.00°C neat) | |
LDLDJEAVRNAEBW-SCSAIBSYSA-N | |
methyl (3R)-3-hydroxybutanoate | |
2724279 | |
97% |
97.0 | |
97% | |
C5H10O3 | |
MFCD00063289 | |
methyl r---3-hydroxybutyrate, r-methyl 3-hydroxybutanoate, methyl 3r-3-hydroxybutanoate, methyl r-3-hydroxybutyrate, r-3-hydroxybutyric acid methyl ester, methyl r-3-hydroxybutanoate, r---3-hydroxybutyric acid methyl ester, r---methyl 3-hydroxybutyrate, r---3-hydroxy-n-butyric acid methyl ester | |
CC(CC(=O)OC)O | |
118.132 | |
118.13 | |
Methyl 3-hydroxybutanoate |